Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045402.1 | NP_001038867.1 | 5 | 846 | 150 | |
XM_005173769.3 | XP_005173826.1 | 5 | 1023 | 150 | ||
GenBank mRNA | BC122267.1 | - | 5 | 846 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |