Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083840.1 | NP_001077309.1 | 6 | 2246 | 125 | |
GenBank mRNA | BC134944.1 | - | 6 | 2246 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |