Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh2ab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008483.2 | NP_001008483.2 | 1 | 843 | 90 | |
GenBank mRNA | AB075933.1 | - | 1 | 843 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Danio rerio Inventoried Both primers and probe map within a single exon |