Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh1g18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024117.2 | NP_001019288.2 | 1 | 757 | 87 | |
GenBank mRNA | AY576943.1 | - | 1 | 757 | 87 | |
BC107998.1 | - | 1 | 946 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |