Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002711.2 | NP_001002711.2 | 4 - 5 | 425 | 86 | |
GenBank mRNA | BC076092.1 | - | 4 - 5 | 426 | 86 | |
EH445440.1 | - | 4 - 5 | 434 | 86 | ||
EH467793.1 | - | 1 - 2 | 524 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |