Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pak2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002717.1 | NP_001002717.1 | 12 - 13 | 1449 | 98 | |
XM_005171262.4 | XP_005171319.1 | 12 - 13 | 1772 | 98 | ||
XM_005171263.4 | XP_005171320.1 | 13 - 14 | 1773 | 98 | ||
GenBank mRNA | BC076511.1 | - | 12 - 13 | 1449 | 98 | |
BC165251.1 | - | 11 - 12 | 1364 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |