Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003539.1 | NP_001003539.1 | 3 - 4 | 531 | 77 | |
GenBank mRNA | BC078187.1 | - | 3 - 4 | 531 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |