Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003753.1 | NP_001003753.1 | 5 - 6 | 763 | 104 | |
GenBank mRNA | BC077106.1 | - | 5 - 6 | 763 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |