Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003890.3 | NP_001003890.3 | 7 - 8 | 1442 | 65 | |
GenBank mRNA | AY648752.1 | - | 7 - 8 | 1401 | 65 | |
BC098545.1 | - | 7 - 8 | 1442 | 65 | ||
HM210100.1 | - | 7 - 8 | 1158 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |