Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007051.3 | NP_001007052.3 | 5 - 6 | 1444 | 73 | |
GenBank mRNA | AY534888.1 | - | 5 - 6 | 1185 | 73 | |
BC066463.1 | - | 5 - 6 | 1341 | 73 | ||
BC098600.1 | - | 5 - 6 | 1444 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |