Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007200.1 | NP_001007201.1 | 15 - 16 | 1522 | 85 | |
GenBank mRNA | BC162720.1 | - | 15 - 16 | 1536 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |