Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008628.1 | NP_001008628.1 | 4 - 5 | 1031 | 94 | |
GenBank mRNA | BC086729.1 | - | 4 - 5 | 1031 | 94 | |
BC165767.1 | - | 4 - 5 | 953 | 94 | ||
BC169189.1 | - | 1014 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |