Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: metrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009983.2 | NP_001009983.2 | 3 - 4 | 802 | 56 | |
GenBank mRNA | BC088383.1 | - | 3 - 4 | 801 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |