Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012250.2 | NP_001012250.1 | 2 - 3 | 393 | 70 | |
GenBank mRNA | AY314756.1 | - | 2 - 3 | 315 | 70 | |
BC098604.1 | - | 2 - 3 | 392 | 70 | ||
BC154349.1 | - | 2 - 3 | 393 | 70 | ||
BC171441.1 | - | 2 - 3 | 382 | 70 | ||
BC171443.1 | - | 2 - 3 | 382 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |