Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mybpc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013511.2 | NP_001013529.2 | 15 - 16 | 1512 | 76 | |
XM_017353828.2 | XP_017209317.1 | 14 - 15 | 1577 | 76 | ||
XM_021470159.1 | XP_021325834.1 | 15 - 16 | 1546 | 76 | ||
XM_021470160.1 | XP_021325835.1 | 14 - 15 | 1574 | 76 | ||
XM_021470161.1 | XP_021325836.1 | 14 - 15 | 1524 | 76 | ||
XM_021470162.1 | XP_021325837.1 | 14 - 15 | 1461 | 76 | ||
GenBank mRNA | BC091662.1 | - | 1506 | 76 | ||
EE719585.1 | - | 5 - 6 | 640 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |