Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014371.1 | NP_001014393.1 | 2 - 3 | 356 | 77 | |
XM_005170889.4 | XP_005170946.1 | 2 - 3 | 435 | 77 | ||
GenBank mRNA | BC091789.1 | - | 2 - 3 | 356 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |