Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: angptl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014821.1 | NP_001014821.1 | 3 - 4 | 817 | 113 | |
XM_009299794.3 | XP_009298069.1 | 4 - 5 | 1142 | 113 | ||
GenBank mRNA | AB199649.1 | - | 3 - 4 | 817 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |