Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017674.1 | NP_001017674.1 | 4 - 5 | 421 | 85 | |
GenBank mRNA | BC093332.1 | - | 4 - 5 | 421 | 85 | |
BC164386.1 | - | 3 - 4 | 363 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |