Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017858.2 | NP_001017858.2 | 4 - 5 | 657 | 101 | |
GenBank mRNA | BC092751.1 | - | 4 - 5 | 604 | 101 | |
BC164455.1 | - | 4 - 5 | 571 | 101 | ||
EH440105.1 | - | 4 - 5 | 630 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |