Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grhpra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020525.1 | NP_001018361.1 | 3 - 4 | 404 | 91 | |
GenBank mRNA | BC095040.1 | - | 3 - 4 | 404 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |