Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tma16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC095357.1 | - | 2 - 3 | 217 | 106 | |
BC152117.1 | - | 2 - 3 | 177 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |