Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024415.2 | NP_001019586.1 | 2 - 3 | 171 | 109 | |
GenBank mRNA | BC095631.1 | - | 2 - 3 | 173 | 109 | |
BC154355.1 | - | 2 - 3 | 170 | 109 | ||
BC164758.1 | - | 2 - 3 | 138 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |