Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025497.1 | NP_001020668.1 | 4 - 5 | 561 | 119 | |
GenBank mRNA | BC163760.1 | - | 4 - 5 | 832 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |