Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs7bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030069.1 | NP_001025240.1 | 3 - 4 | 647 | 74 | |
GenBank mRNA | BC096790.1 | - | 3 - 4 | 647 | 74 | |
BC152595.1 | - | 3 - 4 | 625 | 74 | ||
BC165928.1 | - | 3 - 4 | 478 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |