Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: he1.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098188.1 | NP_001091658.1 | 4 - 5 | 449 | 102 | |
GenBank mRNA | AB175620.1 | - | 4 - 5 | 439 | 102 | |
BC139601.1 | - | 4 - 5 | 449 | 102 | ||
BC162603.1 | - | 4 - 5 | 439 | 102 | ||
BC162606.1 | - | 4 - 5 | 439 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |