Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105702.1 | NP_001099172.1 | 2 - 3 | 335 | 125 | |
GenBank mRNA | BC148172.1 | - | 2 - 3 | 335 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |