Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002447.1 | NP_001002447.1 | 2 - 3 | 146 | 147 | |
XM_005156285.3 | XP_005156342.1 | 2 - 3 | 272 | 147 | ||
GenBank mRNA | BC076024.1 | - | 2 - 3 | 146 | 147 | |
BC152175.1 | - | 2 - 3 | 242 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |