Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itgb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037235.1 | NP_001032312.1 | 12 - 13 | 2068 | 61 | |
GenBank mRNA | BC163266.1 | - | 12 - 13 | 2038 | 61 | |
DQ178647.1 | - | 12 - 13 | 2068 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |