Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077151.1 | NP_001070619.1 | 3 - 4 | 612 | 123 | |
GenBank mRNA | BC122284.1 | - | 3 - 4 | 612 | 123 | |
BC154733.1 | - | 3 - 4 | 613 | 123 | ||
BC157374.1 | - | 3 - 4 | 580 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |