Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077158.2 | NP_001070626.2 | 5 - 6 | 813 | 101 | |
GenBank mRNA | BC122154.1 | - | 5 - 6 | 813 | 101 | |
BC152089.1 | - | 4 - 5 | 1157 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |