Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfml3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077251.1 | NP_001070719.1 | 2 - 3 | 472 | 92 | |
GenBank mRNA | BC114287.1 | - | 2 - 3 | 442 | 92 | |
BC124742.1 | - | 2 - 3 | 472 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |