Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081692.2 | NP_001075161.2 | 17 - 18 | 2243 | 97 | |
XM_005160248.4 | XP_005160305.1 | 17 - 18 | 2201 | 97 | ||
XM_009293357.3 | XP_009291632.1 | 17 - 18 | 2829 | 97 | ||
XM_009293365.3 | XP_009291640.1 | 17 - 18 | 2130 | 97 | ||
GenBank mRNA | BC128874.1 | - | 8 - 9 | 1039 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |