Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080081.1 | NP_001073550.1 | 7 - 8 | 838 | 65 | |
XM_009297718.3 | XP_009295993.1 | 7 - 8 | 898 | 65 | ||
XM_021470533.1 | XP_021326208.1 | 3 - 4 | 206 | 65 | ||
GenBank mRNA | BC128877.1 | - | 7 - 8 | 838 | 65 |
Gene Symbol: LOC110437806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021471060.1 | XP_021326735.1 | 2 - 3 | 137 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |