Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anpepb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089325.1 | NP_001082794.1 | 2 - 3 | 773 | 83 | |
GenBank mRNA | BC115260.1 | - | 2 - 3 | 773 | 83 | |
BC154244.1 | - | 2 - 3 | 774 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |