Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifnphi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111083.1 | NP_001104553.1 | 4 - 5 | 449 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |