Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: opn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111164.1 | NP_001104634.1 | 2 - 3 | 652 | 105 | |
GenBank mRNA | EF043381.1 | - | 2 - 3 | 652 | 105 | |
KT008413.1 | - | 2 - 3 | 646 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Danio rerio Probe spans exons |