Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shrprbck1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111200.1 | NP_001104670.1 | 12 - 13 | 2127 | 64 | |
GenBank mRNA | BC155331.1 | - | 12 - 13 | 2127 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |