Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lifrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113732.1 | NP_001107204.1 | 1 - 2 | 131 | 77 | |
XM_009295324.3 | XP_009293599.1 | 1 - 2 | 174 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |