Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114411.1 | NP_001107883.1 | 2 - 3 | 388 | 143 | |
GenBank mRNA | BC154544.1 | - | 2 - 3 | 388 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |