Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc26a6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114417.1 | NP_001107889.1 | 4 - 5 | 629 | 61 | |
GenBank mRNA | BC155340.1 | - | 4 - 5 | 629 | 61 | |
FJ170818.1 | - | 4 - 5 | 628 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |