Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114424.2 | NP_001107896.2 | 18 - 19 | 2654 | 81 | |
GenBank mRNA | BC157385.1 | - | 18 - 19 | 2613 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |