Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114480.1 | NP_001107952.1 | 3 - 4 | 310 | 90 | |
GenBank mRNA | AM901598.1 | - | 3 - 4 | 407 | 90 | |
BC155620.1 | - | 3 - 4 | 310 | 90 | ||
KM655659.1 | - | 3 - 4 | 212 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |