Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309822.1 | NP_001296751.1 | 8 - 9 | 1085 | 58 | |
NM_131266.2 | NP_571341.2 | 8 - 9 | 1085 | 58 | ||
GenBank mRNA | AF191559.1 | - | 8 - 9 | 1016 | 58 | |
BC059803.1 | - | 8 - 9 | 940 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |