Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msx2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131276.3 | NP_571351.3 | 2 - 3 | 549 | 131 | |
GenBank mRNA | BC109396.1 | - | 2 - 3 | 555 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |