Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pax6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131304.1 | NP_571379.1 | 8 - 9 | 1258 | 75 | |
XM_009297878.3 | XP_009296153.1 | 9 - 10 | 1317 | 75 | ||
XM_009297882.3 | XP_009296157.1 | 10 - 11 | 1389 | 75 | ||
XM_009297883.3 | XP_009296158.1 | 7 - 8 | 1250 | 75 | ||
XM_009297884.3 | XP_009296159.1 | 9 - 10 | 1365 | 75 | ||
XM_009297885.3 | XP_009296160.1 | 8 - 9 | 1322 | 75 | ||
XM_009297886.3 | XP_009296161.1 | 8 - 9 | 1059 | 75 | ||
XM_009297887.3 | XP_009296162.1 | 5 - 6 | 721 | 75 | ||
XM_009297888.3 | XP_009296163.1 | 6 - 7 | 1282 | 75 | ||
XM_009297889.3 | XP_009296164.1 | 5 - 6 | 1240 | 75 | ||
XM_017354048.2 | XP_017209537.1 | 7 - 8 | 1057 | 75 | ||
XM_017354050.2 | XP_017209539.1 | 8 - 9 | 1274 | 75 | ||
XM_021470446.1 | XP_021326121.1 | 9 - 10 | 947 | 75 | ||
XM_021470447.1 | XP_021326122.1 | 7 - 8 | 898 | 75 | ||
XM_021470448.1 | XP_021326123.1 | 8 - 9 | 1025 | 75 | ||
GenBank mRNA | BC066722.1 | - | 7 - 8 | 1181 | 75 | |
X61389.1 | - | 1193 | 75 | |||
X63183.1 | - | 1258 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |