Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131379.2 | NP_571454.2 | 2 - 3 | 237 | 75 | |
GenBank mRNA | AF163311.1 | - | 2 - 3 | 243 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |