Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoxb5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131537.3 | NP_571612.2 | 1 - 2 | 675 | 65 | |
GenBank mRNA | BC078285.1 | - | 1 - 2 | 679 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |