Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126448.2 | NP_001119920.2 | 4 - 5 | 519 | 100 | |
XM_009305008.3 | XP_009303283.1 | 4 - 5 | 467 | 100 | ||
GenBank mRNA | AF322374.1 | - | 4 - 5 | 519 | 100 | |
AI477790.1 | - | 4 - 5 | 337 | 100 | ||
BC066426.1 | - | 4 - 5 | 353 | 100 | ||
BQ826523.1 | - | 4 - 5 | 338 | 100 | ||
EH585281.1 | - | 4 - 5 | 428 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |