Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175576.2 | NP_783166.1 | 2 - 3 | 227 | 72 | |
GenBank mRNA | AY070259.1 | - | 2 - 3 | 227 | 72 | |
BC095297.1 | - | 2 - 3 | 229 | 72 | ||
BC165098.1 | - | 1 - 2 | 220 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |