Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178219.2 | NP_835737.1 | 1 - 2 | 144 | 58 | |
GenBank mRNA | AY182171.1 | - | 1 - 2 | 144 | 58 | |
BC162282.1 | - | 1 - 2 | 95 | 58 | ||
BC162831.1 | - | 1 - 2 | 95 | 58 | ||
KM655731.1 | - | 1 - 2 | 72 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |